6-Methylpyridazine-3-carbonitrile
Supplier: Apollo Scientific
OR929423-1G
OR929423-5G
APOSOR929423-1GEA
613
GBP
APOSOR929423-1G
APOSOR929423-5G
6-Methylpyridazine-3-carbonitrile
6-Methylpyridazine-3-carbonitrile
Formula:
C₆H₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD13190728 CAS Number: 49840-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...