4-Amino-2-hydroxypyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Aminopyridin-2(1H)-one, 4-Amino-1H-pyridin-2-one
OR929418-10G
OR929418-1G
OR929418-5G
APOSOR929418-10GEA
241
GBP
APOSOR929418-10G
APOSOR929418-1G
APOSOR929418-5G
4-Amino-2-hydroxypyridine
4-Amino-2-hydroxypyridine
Formula:
C₅H₆N₂O MW: 110,12 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02682913 CAS Number: 38767-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...