2-Ethynylthiophene 95+%

Supplier: Apollo Scientific
Warning

OR9291-1G OR9291-5G
APOSOR9291-1GEA 19.7 GBP
APOSOR9291-1G APOSOR9291-5G
2-Ethynylthiophene 95+%
2-Ethynylthiophene
Formula: C₆H₄S
MW: 108,1610 g/mol
Boiling Pt: 78…80 °C/88mmHg
Density: 1,08 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD03425129
CAS Number: 4298-52-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR