2-Bromo-2-cyano-N,N-dimethylacetamide 95%
Supplier: Apollo Scientific
Synonyms:
N,N-Dimethyl-2-bromo-2-cyanoacetamide, 2-(N,N-Dimethylcarbamoyl)-2-bromoacetonitrile
OR929093-1G
OR929093-5G
APOSOR929093-1GEA
185
GBP
APOSOR929093-1G
APOSOR929093-5G
2-Bromo-2-cyano-N,N-dimethylacetamide 95%
2-Bromo-2-cyano-N,N-dimethylacetamide
Formula:
C₅H₇BrN₂O MW: 191,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00040509 CAS Number: 15430-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...