Isobutyrophenone 98%

Supplier: Apollo Scientific

Synonyms: Isopropyl phenyl ketone, 2-Methyl-1-phenyl-1-propanone, Phenyl isopropyl ketone, α-Methylpropiophenone

OR929001-100G OR929001-25G OR929001-500G
APOSOR929001-100GEA 103 GBP
APOSOR929001-100G APOSOR929001-25G APOSOR929001-500G
Isobutyrophenone 98%
Isobutyrophenone
Formula: (C₆H₅)C(O)CH(CH₃)₂
MW: 148,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008917
CAS Number: 611-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR