3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid 95%

Supplier: Apollo Scientific

OR928995-1G OR928995-250MG
APOSOR928995-1GEA 649 GBP
APOSOR928995-1G APOSOR928995-250MG
3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid 95%
3-(1,3-Dioxolan-2-yl)cyclobutanecarboxylic acid
Formula: C₈H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD22575225
CAS Number: 1414513-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR