7-Methyl-1H-indazole 98%

Supplier: Apollo Scientific

OR928851-10G OR928851-1G OR928851-5G
APOSOR928851-10GEA 357 GBP
APOSOR928851-10G APOSOR928851-1G APOSOR928851-5G
7-Methyl-1H-indazole 98%
7-Methyl-1H-indazole
Formula: C₈H₈N₂
MW: 132,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663434
CAS Number: 3176-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR