1-Ethyl-1H-indole-5-carbaldehyde 95%

Supplier: Apollo Scientific

OR928526-1G OR928526-5G
APOSOR928526-1GEA 115 GBP
APOSOR928526-1G APOSOR928526-5G
1-Ethyl-1H-indole-5-carbaldehyde 95%
1-Ethyl-1H-indole-5-carbaldehyde
Formula: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD09455244
CAS Number: 944893-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR