2-Methylbenzofuran 95%
Supplier: Apollo Scientific
OR928246-1G
OR928246-250MG
APOSOR928246-1GEA
296
GBP
APOSOR928246-1G
APOSOR928246-250MG
2-Methylbenzofuran 95%
2-Methylbenzofuran
Formula:
C₉H₈O MW: 132,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005850 CAS Number: 4265-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...