4-Methylthiopiperidine hydrochloride 95%

Supplier: Apollo Scientific

OR928219-1G OR928219-5G
APOSOR928219-1GEA 223 GBP
APOSOR928219-1G APOSOR928219-5G
4-Methylthiopiperidine hydrochloride 95%
4-Methylthiopiperidine hydrochloride
Formula: C₆H₁₄ClNS
Storage Temperature: Ambient
MDL Number: MFCD13188535
CAS Number: 208245-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR