N,N-Dimethyl-1,1,1-trimethylsilylamine 95%

Supplier: Apollo Scientific
Danger

Synonyms: (Dimethylamino)trimethylsilane, Trimethylsilyldimethylamine, N,N-Dimethyltrimethylsilylamine, TMSDMA, N-(Trimethylsilyl)dimethylamine, N,N-Dimethylaminotrimethylsilane, Pentamethylsilylamine

OR927800-100G OR927800-25G
APOSOR927800-100GEA 591 GBP
APOSOR927800-100G APOSOR927800-25G
N,N-Dimethyl-1,1,1-trimethylsilylamine 95%
N,N-Dimethyl-1,1,1-trimethylsilylamine
Formula: C₅H₁₅NSi
MW: 117,27 g/mol
Boiling Pt: 86 °C
Density: 0,732 g/cm³ (25 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00008297
CAS Number: 2083-91-2
UN: 2733
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR