3-Benzyloxypyridine 95%

Supplier: Apollo Scientific

OR927333-1G OR927333-250MG
APOSOR927333-1GEA 728 GBP
APOSOR927333-1G APOSOR927333-250MG
3-Benzyloxypyridine 95%
3-Benzyloxypyridine
Formula: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD13151998
CAS Number: 76509-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR