6-Chloro-1,2,3,4-tetrahydroisoquinoline 97%
Supplier: Apollo Scientific
OR926186-1G
OR926186-250MG
APOSOR926186-1GEA
342
GBP
APOSOR926186-1G
APOSOR926186-250MG
6-Chloro-1,2,3,4-tetrahydroisoquinoline 97%
6-Chloro-1,2,3,4-tetrahydroisoquinoline
Formula:
C₉H₁₀ClN MW: 167,6354 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06739156 CAS Number: 33537-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...