2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one 95%
Supplier: Apollo Scientific
OR925640-1G
OR925640-5G
APOSOR925640-1GEA
205
GBP
APOSOR925640-1G
APOSOR925640-5G
2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one 95%
2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one
Formula:
C₁₀H₁₃N₃O Storage Temperature: Ambient |
MDL Number:
MFCD00520824 CAS Number: 21599-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...