2-Chloroquinolin-4-amine 95%

Supplier: Apollo Scientific

OR925609-1G OR925609-250MG
APOSOR925609-1GEA 91.8 GBP
APOSOR925609-1G APOSOR925609-250MG
2-Chloroquinolin-4-amine 95%
2-Chloroquinolin-4-amine
Formula: C₉H₇ClN₂
MW: 178,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705643
CAS Number: 80947-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR