Diacetone acrylamide 95%

Supplier: Apollo Scientific

Synonyms: N-(2-Methyl-4-oxopentan-2-yl)acrylamide, N-(1,1-Dimethyl-3-oxobutyl)acrylamide

OR925264-25G OR925264-100G OR925264-500G
APOSOR925264-25GEA 106 GBP
APOSOR925264-25G APOSOR925264-100G APOSOR925264-500G
Diacetone acrylamide 95%
Diacetone acrylamide
Formula: C₉H₁₅NO₂
MW: 169,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008788
CAS Number: 2873-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR