4-Ethylpyrocatechol 97%
Supplier: Apollo Scientific
Synonyms:
4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
OR925237-10G
OR925237-1G
OR925237-5G
APOSOR925237-10GEA
302
GBP
APOSOR925237-10G
APOSOR925237-1G
APOSOR925237-5G
4-Ethylpyrocatechol 97%
4-Ethylpyrocatechol
Formula:
C₈H₁₀O₂ MW: 138,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015847 CAS Number: 1124-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...