Potassium bis(1-pyrazolyl)borohydride 95%

Supplier: Apollo Scientific

Synonyms: Potassium Dihydrobis(1-pyrazolyl)borate

OR925156-1G OR925156-5G
APOSOR925156-1GEA 197 GBP
APOSOR925156-1G APOSOR925156-5G
Potassium bis(1-pyrazolyl)borohydride 95%
Potassium bis(1-pyrazolyl)borohydride
Formula: C₆H₈BKN₄
MW: 186,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058739
CAS Number: 18583-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR