4-Methylhippuric acid 95%
Supplier: Apollo Scientific
OR925154-25G
OR925154-5G
APOSOR925154-25GEA
121
GBP
APOSOR925154-25G
APOSOR925154-5G
4-Methylhippuric acid 95%
4-Methylhippuric acid
Formula:
C₁₀H₁₁NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00020449 CAS Number: 27115-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...