N,N-Di-n-propylformamide 95%

Supplier: Apollo Scientific

OR925014-25G OR925014-5G
APOSOR925014-25GEA 89.1 GBP
APOSOR925014-25G APOSOR925014-5G
N,N-Di-n-propylformamide 95%
N,N-Di-n-propylformamide
Formula: HCON(CH₂CH₂CH₃)₂
MDL Number: MFCD00043518
CAS Number: 6282-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR