1-Ethyl-1-(m-tolyl)hydrazine 95%

Supplier: Apollo Scientific

OR924958-1G OR924958-25G OR924958-5G
APOSOR924958-1GEA 145 GBP
APOSOR924958-1G APOSOR924958-25G APOSOR924958-5G
1-Ethyl-1-(m-tolyl)hydrazine 95%
1-Ethyl-1-(m-tolyl)hydrazine
Formula: C₉H₁₄N₂
MW: 150,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844792
CAS Number: 99717-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR