1-Acetyl-1,2,3,4-tetrahydroquinoline 98%

Supplier: Apollo Scientific

Synonyms: 1,2,3,4-Tetrahydro-1-acetylquinoline

OR924756-100G OR924756-25G OR924756-5G
APOSOR924756-100GEA 584 GBP
APOSOR924756-100G APOSOR924756-25G APOSOR924756-5G
1-Acetyl-1,2,3,4-tetrahydroquinoline 98%
1-Acetyl-1,2,3,4-tetrahydroquinoline
Formula: C₁₁H₁₃NO
MW: 175,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01195305
CAS Number: 4169-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR