2-Isopropylimidazole 98%
Supplier: Apollo Scientific
Synonyms:
2-Isopropyl-1H-imidazole
OR924662-100G
OR924662-1KG
OR924662-500G
APOSOR924662-100GEA
55.1
GBP
APOSOR924662-100G
APOSOR924662-1KG
APOSOR924662-500G
2-Isopropylimidazole 98%
2-Isopropylimidazole
Formula:
C₆H₁₀N₂ MW: 110,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014486 CAS Number: 36947-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...