3-Iodo-1-propanol 95%

Supplier: Apollo Scientific

OR924612-1G OR924612-5G
APOSOR924612-1GEA 112 GBP
APOSOR924612-1G APOSOR924612-5G
3-Iodo-1-propanol 95%
3-Iodo-1-propanol
Formula: C₃H₇IO
MW: 185,99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060329
CAS Number: 627-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR