Phthalic acid 98%

Supplier: Apollo Scientific

Synonyms: Pathalic acid

OR923932-100G OR923932-1KG OR923932-500G
APOSOR923932-100GEA 47.2 GBP
APOSOR923932-100G APOSOR923932-1KG APOSOR923932-500G
Phthalic acid 98%
Phthalic acid
Formula: C₈H₆O₄
MW: 166,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002467
CAS Number: 88-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR