2-Methoxy-6-(methylamino)pyridine 95%

Supplier: Apollo Scientific

OR923913-25G OR923913-5G
APOSOR923913-25GEA 108 GBP
APOSOR923913-25G APOSOR923913-5G
2-Methoxy-6-(methylamino)pyridine 95%
2-Methoxy-6-(methylamino)pyridine
Formula: C₇H₁₀N₂O
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191557
CAS Number: 88569-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR