3'-(Dimethylamino)acetophenone 95%

Supplier: Apollo Scientific

OR923846-1G OR923846-5G
APOSOR923846-1GEA 119 GBP
APOSOR923846-1G APOSOR923846-5G
3'-(Dimethylamino)acetophenone 95%
3'-(Dimethylamino)acetophenone
Formula: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00059343
CAS Number: 18992-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR