1-Methylpyrrolidine-2-thione 96%
Supplier: Apollo Scientific
OR923785-1G
OR923785-5G
APOSOR923785-1GEA
106
GBP
APOSOR923785-1G
APOSOR923785-5G
1-Methylpyrrolidine-2-thione 96%
1-Methylpyrrolidine-2-thione
Formula:
C₅H₉NS MW: 115,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00966332 CAS Number: 10441-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...