4-Methyl-4-heptanol 95%

Supplier: Apollo Scientific

OR923726-1G OR923726-5G
APOSOR923726-1GEA 119 GBP
APOSOR923726-1G APOSOR923726-5G
4-Methyl-4-heptanol 95%
4-Methyl-4-heptanol
Formula: C₈H₁₈O
MW: 130,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021840
CAS Number: 598-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR