1-Isopropylpyrrolidin-3-ylamine dihydrochloride 95%
Supplier: Apollo Scientific
Synonyms:
1-Isopropyl-pyrrolidin-3-ylamine dihydrochloride, 1-(Propan-2-yl)pyrrolidin-3-amine
OR923288-1G
OR923288-5G
APOSOR923288-1GEA
114
GBP
APOSOR923288-1G
APOSOR923288-5G
1-Isopropylpyrrolidin-3-ylamine dihydrochloride 95%
1-Isopropylpyrrolidin-3-ylamine dihydrochloride
Formula:
C₇H₁₈Cl₂N₂ MW: 201,1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08544449 CAS Number: 19985-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...