3-Phenylpropanal 95%

Supplier: Apollo Scientific

Synonyms: 3-Phenylpropionaldehyde, Hydrocinnamaldehyde

OR923069-100G OR923069-1KG OR923069-500G
APOSOR923069-100GEA 119 GBP
APOSOR923069-100G APOSOR923069-1KG APOSOR923069-500G
3-Phenylpropanal 95%
3-Phenylpropanal
Formula: C₉H₁₀O
MW: 134,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007021
CAS Number: 104-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR