N-(2-Hydroxyethyl)phthalimide 98%
Supplier: Apollo Scientific
Synonyms:
2-(2-Hydroxyethyl)isoindoline-1,3-dione
OR923052-100G
OR923052-1KG
OR923052-500G
APOSOR923052-100GEA
48.6
GBP
APOSOR923052-100G
APOSOR923052-1KG
APOSOR923052-500G
N-(2-Hydroxyethyl)phthalimide 98%
N-(2-Hydroxyethyl)phthalimide
Formula:
C₁₀H₉NO₃ MW: 191,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005903 CAS Number: 3891-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...