2,4-Dimethoxyphenol 97%
Supplier: Apollo Scientific
OR922810-1G
OR922810-5G
APOSOR922810-1GEA
237
GBP
APOSOR922810-1G
APOSOR922810-5G
2,4-Dimethoxyphenol 97%
2,4-Dimethoxyphenol
Formula:
C₈H₁₀O₃ MW: 154,1632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060637 CAS Number: 13330-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...