6-Methyl-2-heptanone 95%

Supplier: Apollo Scientific

Synonyms: Methyl 4-Methylpentyl ketone, 6-Methylheptan-2-one

OR922705-10G OR922705-1G OR922705-5G
APOSOR922705-10GEA 368 GBP
APOSOR922705-10G APOSOR922705-1G APOSOR922705-5G
6-Methyl-2-heptanone 95%
6-Methyl-2-heptanone
Formula: C₈H₁₆O
MW: 128,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026527
CAS Number: 928-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR