ε-Hexalactone 95%

Supplier: Apollo Scientific

Synonyms: ε-Caprolactone, Oxepan-2-one, Hexan-6-olide

OR922393-100G OR922393-1KG OR922393-500G
APOSOR922393-100GEA 98.3 GBP
APOSOR922393-100G APOSOR922393-1KG APOSOR922393-500G
ε-Hexalactone 95%
ε-Hexalactone
Formula: C₆H₁₀O₂
MW: 114,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003267
CAS Number: 502-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR