Diisobutyl ether 95%

Supplier: Apollo Scientific

Synonyms: Isobutyl Ether

OR921593-25G OR921593-5G
APOSOR921593-25GEA 0 GBP
APOSOR921593-25G APOSOR921593-5G
Diisobutyl ether 95%
Diisobutyl ether
Formula: C₈H₁₈O
MW: 130,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008935
CAS Number: 628-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR