9-Oxabicyclo[6.1.0]-4-nonene 98%
Supplier: Apollo Scientific
OR920820-1G
OR920820-25G
OR920820-5G
APOSOR920820-1GEA
139
GBP
APOSOR920820-1G
APOSOR920820-25G
APOSOR920820-5G
9-Oxabicyclo[6.1.0]-4-nonene 98%
9-Oxabicyclo[6.1.0]-4-nonene
Formula:
C₈H₁₂O MW: 124,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00086420 CAS Number: 637-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...