2-Methyl-3-isopropoxypyrazine 95%

Supplier: Apollo Scientific

Synonyms: 2-Isopropoxy-3-methylpyrazine

OR920655-1G OR920655-5G
APOSOR920655-1GEA 157 GBP
APOSOR920655-1G APOSOR920655-5G
2-Methyl-3-isopropoxypyrazine 95%
2-Methyl-3-isopropoxypyrazine
Formula: C₈H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00059772
CAS Number: 94089-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR