N-(3-Aminopropyl)-N-methylaniline 95%
Supplier: Apollo Scientific
Synonyms:
N1-Methyl-N1-phenylpropane-1,3-diamine
OR920279-25G
OR920279-100G
APOSOR920279-25GEA
171
GBP
APOSOR920279-25G
APOSOR920279-100G
N-(3-Aminopropyl)-N-methylaniline 95%
N-(3-Aminopropyl)-N-methylaniline
Formula:
C₁₀H₁₆N₂ MW: 164,250 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191602 CAS Number: 53485-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...