2-Benzyloxy-1,3-propanediol 95%
Supplier: Apollo Scientific
Synonyms:
2-(Benzyloxy)propane-1,3-diol, 2-O-Benzylglycerol, Glycerol 2-Benzyl Ether
OR920067-1G
OR920067-250MG
APOSOR920067-1GEA
179
GBP
APOSOR920067-1G
APOSOR920067-250MG
2-Benzyloxy-1,3-propanediol 95%
2-Benzyloxy-1,3-propanediol
Formula:
C₁₀H₁₄O₃ MW: 182,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00075493 CAS Number: 14690-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...