1-tert-Butoxy-4-chlorobenzene 95%

Supplier: Apollo Scientific

Synonyms: tert-Butyl 4-Chlorophenyl Ether, 1-(tert-Butoxy)-4-chlorobenzene

OR920002-25G OR920002-5G
APOSOR920002-25GEA 97.1 GBP
APOSOR920002-25G APOSOR920002-5G
1-tert-Butoxy-4-chlorobenzene 95%
1-tert-Butoxy-4-chlorobenzene
Formula: C₁₀H₁₃ClO
MW: 184,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038411
CAS Number: 18995-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR