2,4,4-Trimethyl-2-oxazoline 98%
Supplier: Apollo Scientific
Synonyms:
2,4,4-Trimethyl-4,5-dihydrooxazole
OR919620-25G
OR919620-5G
APOSOR919620-25GEA
328
GBP
APOSOR919620-25G
APOSOR919620-5G
2,4,4-Trimethyl-2-oxazoline 98%
2,4,4-Trimethyl-2-oxazoline
Formula:
C₆H₁₁NO MW: 113,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005301 CAS Number: 1772-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...