Phenyl vinyl sulphoxide 95%

Supplier: Apollo Scientific

Synonyms: (Vinylsulphinyl)benzene

OR919122-25G OR919122-5G
APOSOR919122-25GEA 728 GBP
APOSOR919122-25G APOSOR919122-5G
Phenyl vinyl sulphoxide 95%
Phenyl vinyl sulphoxide
Formula: C₈H₈OS
MW: 152,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002086
CAS Number: 20451-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR