4-Methylthiophene-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR919078-1G
OR919078-250MG
APOSOR919078-1GEA
155
GBP
APOSOR919078-1G
APOSOR919078-250MG
4-Methylthiophene-3-carboxylic acid 95%
4-Methylthiophene-3-carboxylic acid
Formula:
C₆H₆O₂S MW: 142,1756 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02257717 CAS Number: 78071-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...