Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate 98%

Supplier: Apollo Scientific

Synonyms: (R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid Methyl Ester, (R)-Methyl 2-(4-hydroxyphenoxy)propanoate

OR919073-25G OR919073-5G
APOSOR919073-25GEA 178 GBP
APOSOR919073-25G APOSOR919073-5G
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate 98%
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate
Formula: C₁₀H₁₂O₄
MW: 196,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00274087
CAS Number: 96562-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR