Benzyloxyacetyl chloride 96%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Benzyloxy)acetyl chloride

OR919060-10G OR919060-25G
APOSOR919060-10GEA 165 GBP
APOSOR919060-10G APOSOR919060-25G
Benzyloxyacetyl chloride 96%
Benzyloxyacetyl chloride
Formula: C₉H₉ClO₂
MW: 184,62 g/mol
Boiling Pt: 105…107 °C/5mmHg
Density: 1,17 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010768
CAS Number: 19810-31-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR