3-Bromotetrahydro-2H-pyran 96%

Supplier: Apollo Scientific

Synonyms: 3-Bromo-tetrahydropyran

OR918100-1G OR918100-250MG
APOSOR918100-1GEA 178 GBP
APOSOR918100-1G APOSOR918100-250MG
3-Bromotetrahydro-2H-pyran 96%
3-Bromotetrahydro-2H-pyran
Formula: C₅H₉BrO
MW: 165,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371502
CAS Number: 13047-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR