Ethyl-1-methylpyrazole-4-carboxylate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 1-methyl-1H-pyrazole-4-carboxylate
OR917786-1G
OR917786-5G
APOSOR917786-1GEA
205
GBP
APOSOR917786-1G
APOSOR917786-5G
Ethyl-1-methylpyrazole-4-carboxylate 95%
Ethyl-1-methylpyrazole-4-carboxylate
Formula:
C₇H₁₀N₂O₂ MW: 154,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12180234 CAS Number: 85290-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...