[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,6-Dimethylanilino(oxo)acetic acid, 2-((2,6-Dimethylphenyl)amino)-2-oxoacetic acid, N-(2,6-Dimethylphenyl)oxamic acid, DMPAO, (2,6-Dimethylanilino)(oxo)acetic acid, [(2,6-Dimethylphenyl)Carbamoyl]formic acid
OR917428-1G
OR917428-5G
APOSOR917428-1GEA
159
GBP
APOSOR917428-1G
APOSOR917428-5G
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid 98%
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid
Formula:
C₁₀H₁₁NO₃ MW: 193,20 g/mol Melting Pt: 173 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD10018453 CAS Number: 2903-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...