Methyl butyrylacetate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 3-oxohexanoate

OR917137-100G OR917137-500G OR917137-25G
APOSOR917137-100GEA 89.2 GBP
APOSOR917137-100G APOSOR917137-500G APOSOR917137-25G
Methyl butyrylacetate 95%
Methyl butyrylacetate
Formula: C₇H₁₂O₃
MW: 144,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040995
CAS Number: 30414-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR